Learn More
2-Bromo-3-(bromomethyl)pyridine, 96%, Thermo Scientific Chemicals
Product Code. 15468947
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15468947 1 g 1g
15478947 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15468947 Supplier Thermo Scientific Alfa Aesar Supplier No. H52311.03
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
CAS: 94446-97-6 | Molecular Formula: C6H5Br2N | Molecular Weight: 250.921 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 94446-97-6
Molecular Formula C6H5Br2N
Molecular Weight (g/mol) 250.921
MDL Number MFCD08276299
InChI Key NEWLOSCWMVKPGI-UHFFFAOYSA-N
Synonym 2-bromo-3-bromomethyl pyridine, 2-bromo-3-bromomethylene pyridine, pubchem22284, 2-bromo-3-bromomethyl-pyridine, pyridine, 2-bromo-3-bromomethyl
PubChem CID 10901078
IUPAC Name 2-bromo-3-(bromomethyl)pyridine
SMILES C1=CC(=C(N=C1)Br)CBr
Specifications
Melting Point 38°C
Boiling Point 291°C
Flash Point >110°C (230°F)
Quantity 1 g
Formula Weight 250.92
Percent Purity 96%
Chemical Name or Material 2-Bromo-3-(bromomethyl)pyridine