Learn More
2-Bromo-2'-methoxyacetophenone, 98%, Thermo Scientific Chemicals
Catalog Number p-7044674
Click to view available options
Quantity:
1 g
5 g
31949-21-0
C9H9BrO2
229.073
MFCD00000196
GKNCPTLOPRDYMH-UHFFFAOYSA-N
2-bromo-1-2-methoxyphenyl ethanone, 2-bromo-2'-methoxyacetophenone, 2-methoxyphenacyl bromide, o-methoxy phenacylbromide, bromomethyl 2-methoxyphenyl ketone, 2-bromo-1-2-methoxyphenyl ethan-1-one, 2-bromo-2'-methoxy acetophenone, ethanone, 2-bromo-1-methoxyphenyl, alpha-bromo-o-methoxyacetophenone, 2-bromoacetylanisole
123440
2-bromo-1-(2-methoxyphenyl)ethanone
COC1=CC=CC=C1C(=O)CBr
31949-21-0
C9H9BrO2
229.073
MFCD00000196
GKNCPTLOPRDYMH-UHFFFAOYSA-N
2-bromo-1-2-methoxyphenyl ethanone, 2-bromo-2'-methoxyacetophenone, 2-methoxyphenacyl bromide, o-methoxy phenacylbromide, bromomethyl 2-methoxyphenyl ketone, 2-bromo-1-2-methoxyphenyl ethan-1-one, 2-bromo-2'-methoxy acetophenone, ethanone, 2-bromo-1-methoxyphenyl, alpha-bromo-o-methoxyacetophenone, 2-bromoacetylanisole
123440
2-bromo-1-(2-methoxyphenyl)ethanone
COC1=CC=CC=C1C(=O)CBr
2-Bromo-2'-methoxyacetophenone is used as a pharmaceutical intermediate .
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Bromo-2′-methoxyacetophenone is used as a pharmaceutical intermediate .
Solubility
Insoluble in water.
Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 42°C to 46°C
Boiling Point 130°C (1 mmHg)
Flash Point >110°C (230°F)
Quantity 1 g
UN Number UN3261
Beilstein 1938788
Solubility Information Insoluble in water.
Formula Weight 229.08
Percent Purity 98%
Chemical Name or Material 2-Bromo-2'-methoxyacetophenone