Learn More
2-Bromo-1,3-diethylbenzene, 94%
Product Code. 11342176
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11342176 5 g 5g
11332176 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11342176 Supplier Thermo Scientific Alfa Aesar Supplier No. L03482.06
Request Bulk or Custom Format
CAS: 65232-57-7 | C10H13Br | 213.12 g/mol
2-Bromo-1,3-diethylbenzene is used as raw material in synthetic organic chemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Bromo-1,3-diethylbenzene is used as raw material in synthetic organic chemistry.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 65232-57-7
Molecular Formula C10H13Br
Molecular Weight (g/mol) 213.12
MDL Number MFCD00017770
InChI Key FPWGIABDOFXETH-UHFFFAOYSA-N
Synonym 2,6-diethylbromobenzene, 1-bromo-2,6-diethylbenzene, 2,6-diethyl bromobenzene, benzene, 2-bromo-1,3-diethyl, acmc-20ao3d, 2,6-diethylphenylbromide, 2,6-diethylphenyl bromide, 1,3-diethyl-2-bromobenzene, 2-bromo-1,3-diethyl-benzene, #
PubChem CID 522160
IUPAC Name 2-bromo-1,3-diethylbenzene
SMILES CCC1=CC=CC(CC)=C1Br
Specifications
Density 1.264
Boiling Point 78°C to 80°C (2 mmHg)
Refractive Index 1.544
Quantity 5 g
Beilstein 2501180
Solubility Information Insoluble in water.
Formula Weight 213.12
Percent Purity 94%
Chemical Name or Material 2-Bromo-1,3-diethylbenzene