Learn More
2-(Boc-amino)-6-methylpyridine, 97%, Thermo Scientific™
Product Code. 15497467
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15497467 5 g 5g
15487467 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15497467 Supplier Thermo Scientific Alfa Aesar Supplier No. H35121.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 90101-22-7
Molecular Formula C11H16N2O2
Molecular Weight (g/mol) 208.26
MDL Number MFCD07776935
InChI Key XSVAARVWQDEAEL-UHFFFAOYSA-N
Synonym tert-butyl 6-methylpyridin-2-yl carbamate, tert-butyl 6-methylpyridin-2-ylcarbamate, tert-butyl n-6-methylpyridin-2-yl carbamate, 2-boc-amino-6-picoline, 6-methyl-pyridin-2-yl-carbamic acid tert-butyl ester, 2-boc-amino-6-methylpyridine, carbamic acid, 6-methyl-2-pyridinyl-, 1,1-dimethylethyl ester, 2-boc protected, 2-n-boc-amino-6-picoline
PubChem CID 11367776
IUPAC Name tert-butyl N-(6-methylpyridin-2-yl)carbamate
SMILES CC1=NC(NC(=O)OC(C)(C)C)=CC=C1
Specifications
Melting Point 63°C to 64°C
Quantity 5 g
Formula Weight 208.26
Percent Purity 97%
Chemical Name or Material 2-(Boc-amino)-6-methylpyridine