missing translation for 'onlineSavingsMsg'
Learn More

2-(Bismethyl)aminomethylcyclohexanone Hydrochloride, TRC

Product Code. 17924335
Change view
Click to view available options
Quantity:
2.5 g
25 g
Unit Size:
2.5g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17924335 2.5 g 2.5g
30134403 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17924335 Supplier Toronto Research Chemicals Supplier No. TRCB5854452.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42036-65-7
Formula Weight 191.1077
InChI Formula InChI=1S/C9H17NO.ClH/c1-10(2)7-8-5-3-4-6-9(8)11;/h8H,3-7H2,1-2H3;1H
IUPAC Name 2-[(dimethylamino)methyl]cyclohexan-1-one;hydrochloride
Molecular Formula C9 H17 N O . Cl H
Molecular Weight (g/mol) 191.7
SMILES Cl.CN(C)CC1CCCCC1=O
Synonym (2RS)-2-[(Dimethylamino)methyl]cyclohexanone Hydrochloride, Tramadol Hydrochloride Imp. E (EP) as Hydrochloride
Chemical Name or Material (2RS)-2-[(Dimethylamino)methyl]cyclohexanone Hydrochloride

Specifications

Quantity 2.5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.