Learn More
2-Benzylsuccinic acid, 95%, BLD Pharm™
Product Code. 31335389
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31335389 250 mg
31323665 1 g
31326604 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 31335389 Supplier BLD Pharm Supplier No. BD126457250mg
 Restricted Item
Estimated Shipment: 25-11-2026
Specifications
CAS 884-33-3
Molecular Formula C11H12O4
MDL Number MFCD00055798
Quantity 250 mg
SMILES OC(=O)CC(CC1=CC=CC=C1)C(O)=O
Molecular Weight (g/mol) 208.21
Percent Purity 95%
Chemical Name or Material 2-Benzylsuccinic acid