Learn More
2-Benzamido-2-hydroxyacetic acid, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 16555-77-4 |
| Molecular Formula | C9H9NO4 |
| MDL Number | MFCD00004228 |
| Quantity | 25 g |
| SMILES | O=C(O)C(NC(C1=CC=CC=C1)=O)O |
| Molecular Weight (g/mol) | 195.17 |
| Percent Purity | 95% |
| Chemical Name or Material | 2-Benzamido-2-hydroxyacetic acid |