Learn More
2-Azidoethyl 2-Acetamido-2-deoxy-b-D-glucopyranoside, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Azidoethyl 2-Acetamido-2-deoxy-beta-D-glucopyranoside |
| CAS | 142072-12-6 |
| Molecular Formula | C10H18N4O6 |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C10H18N4O6/c1-5(16)13-7-9(18)8(17)6(4-15)20-10(7)19-3-2-12-14-11/h6-10,15,17-18H,2-4H2,1H3,(H,13,16)/t6-,7-,8-,9-,10-/m1/s1 |
| SMILES | CC(=O)N[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]1OCCN=[N+]=[N-] |
| IUPAC Name | N-[(2R,3R,4R,5S,6R)-2-(2-azidoethoxy)-4,5-dihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide |
| Molecular Weight (g/mol) | 290.28 |
| Formula Weight | 290.1226 |