Learn More
2-Aminophenol, TRC
Product Code. 30320963
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30320963 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30320963 Supplier Toronto Research Chemicals Supplier No. TRCA6189305G
This item requires a price update prior to being ordered.
Request a Price for this item here.
, 02 950 59 478
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Aminophenol
CAS 95-55-6
Molecular Formula C6 H7 N O
Quantity 5 g
Synonym Phenol, o-amino- (8CI), 2-Aminophenol, (2-Hydroxyphenyl)amine, 1-Amino-2-hydroxybenzene, 1-Hydroxy-2-aminobenzene, 2-Amino-1-hydroxybenzene, 2-Aminophenyl alcohol, 2-Hydroxyaniline, 2-Hydroxybenzenamine, BASF Ursol 3GA, Benzofur GG, C.I. 76520, C.I. Oxidation Base 17, Fouramine OP, NSC 1534, NSC 226261, Nako Yellow 3GA, Paradone Olive Green B, Pelagol 3GA, Pelagol Grey GG, Rodol 2G, Zoba 3GA, o-Aminohydroxybenzene, o-Aminophenol, o-Hydroxyaniline, o-Hydroxyphenylamine
InChI Formula InChI=1S/C6H7NO/c7-5-3-1-2-4-6(5)8/h1-4,8H,7H2
SMILES Nc1ccccc1O
IUPAC Name 2-aminophenol
Molecular Weight (g/mol) 109.13
Formula Weight 109.0528