missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoindan-2-carboxylic Acid Hydrochloride, 97%, Thermo Scientific™

Product Code. 10799694 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Glass bottle
Unit Size:
250mg
Product Code. Quantity Packaging unitSize
10799694 250 mg Glass bottle 250mg
1 options
This item is not returnable. View return policy

Product Code. 10799694

Brand: Thermo Scientific Acros 376382500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 33584-60-0
CAS Min % 96.0
CAS Max % 100.0
Melting Point 285°C to 289°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Molecular Formula C10H11NO2·ClH
MDL Number MFCD00800587
Quantity 250 mg
Synonym 2-amino-2,3-dihydro-1h-indene-2-carboxylic acid hydrochloride, 2-aminoindan-2-carboxylic acid hydrochloride, 2-amino-1,3-dihydroindene-2-carboxylic acid hydrochloride, 1h-indene-2-carboxylicacid, 2-amino-2,3-dihydro-, hydrochloride 1:1
InChI Key TUBGNZQKMUMUEA-UHFFFAOYSA-N
SMILES C1C2=CC=CC=C2CC1(C(=O)O)N.Cl
IUPAC Name 2-amino-1,3-dihydroindene-2-carboxylic acid;hydrochloride
Molecular Weight (g/mol) 213.66
PubChem CID 45789447
Formula Weight 213.66
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminoindan-2-carboxylic acid hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.