Learn More
2-Aminoethyldiphenyl Borate, Sigma-Aldrich™
Specifications
Specifications
| CAS | 524-95-8 |
| Melting Point | 189°C to 194°C |
| MDL Number | MFCD00014823 |
| Quantity | 5 g |
| Synonym | 2-Aminoethyldiphenyl borate, Diphenylboric acid-2-aminoethyl ester, 2-Aminoethyldiphenyl borate |
| Molecular Weight (g/mol) | 225.09 |
| Grade | Research |