Learn More
2-Aminoethyl Dihydrogen Phosphate, Sigma-Aldrich™
Specifications
Specifications
| CAS | 1071-23-4 |
| Melting Point | 241°C to 243°C |
| Molecular Formula | H2NCH2CH2OPO(OH)2 |
| MDL Number | MFCD00008178 |
| Quantity | 10 g |
| Synonym | 2-Aminoethyl dihydrogen phosphate, Colamine phosphoric acid, Ethanolamine dihydrogen phosphate, 2-Hydroxyethylammonium dihydrogen phosphate, O-Phosphoethanolamine, 2-Aminoethyl dihydrogen phosphate |
| Molecular Weight (g/mol) | 141.06 |
| Grade | Research |