missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzeneboronic acid, 96%

Product Code. p-7047219
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
This item is not returnable. View return policy

Product Code. 11304905

Brand: Thermo Scientific Alfa Aesar L18069.MC

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5570-18-3 | C6H8BNO2 | 136.945 g/mol

2-Aminobenzeneboronic acid derivative is used in the synthesis of quinolines by reacting with alpha, beta unsaturated ketones. Its hydrochloride derivative is used in the preparation of designed boronate ligands for glucose-selective holographic sensors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Aminobenzeneboronic acid derivative is used in the synthesis of quinolines by reacting with alpha, beta unsaturated ketones. Its hydrochloride derivative is used in the preparation of designed boronate ligands for glucose-selective holographic sensors.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5570-18-3
Molecular Formula C6H8BNO2
Molecular Weight (g/mol) 136.945
MDL Number MFCD01074645
InChI Key DIRRKLFMHQUJCM-UHFFFAOYSA-N
Synonym 2-aminophenyl boronic acid, 2-aminobenzeneboronic acid, o-aminophenylboronic acid, 2-aminophenylboronicacid, 2-boronoaniline, pubchem9885, acmc-1apjj, 2-amino-phenylboronic acid, 2-amino benzeneboronic acid
PubChem CID 2773216
IUPAC Name (2-aminophenyl)boronic acid
SMILES B(C1=CC=CC=C1N)(O)O

Specifications

Melting Point 188°C to 190°C
Quantity 100 mg
Beilstein 2962939
Formula Weight 136.95
Percent Purity 96%
Chemical Name or Material 2-Aminobenzeneboronic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.