Learn More
2-Amino-7-bromofluorene, tech. 90%
Product Code. 15461588
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15461588 1 g 1g
15481588 250 mg 250mg
15471588 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15461588 Supplier Thermo Scientific Alfa Aesar Supplier No. H55370.03
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
CAS: 6638-60-4 | C13H10BrN | 260.134 g/mol
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Sparingly soluble in water. (0.724 mg/L)
Notes
Incompatible with oxidizing agents. Store at room temperature.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6638-60-4
Molecular Formula C13H10BrN
Molecular Weight (g/mol) 260.134
MDL Number MFCD00001127
InChI Key RJWYTISHBMNMOZ-UHFFFAOYSA-N
Synonym 2-amino-7-bromofluorene, 2-bromo-7-aminofluorene, fluoren-2-amine, 7-bromo, 7-bromo-9h-fluoren-2-ylamine, 9h-fluoren-2-amine, 7-bromo, 7-bromofluorene-2-ylamine, acmc-20a5iw, 2-amino-7-bromo-9h-fluorene, 9h-fluoren-2-amine,7-bromo, 7-bromanyl-9h-fluoren-2-amine
PubChem CID 23126
IUPAC Name 7-bromo-9H-fluoren-2-amine
SMILES C1C2=C(C=CC(=C2)N)C3=C1C=C(C=C3)Br
Specifications
Melting Point 148°C to 149°C
Quantity 1 g
Solubility Information Sparingly soluble in water. (0.724mg/L)
Formula Weight 260.13
Percent Purity 90%
Grade Technical
Chemical Name or Material 2-Amino-7-bromofluorene