Learn More
2-Amino-6-ethylpyridine, 97%
Product Code. 11429436
Click to view available options
Quantity:
2.5 g
10 g
50 g
Unit Size:
10g
2.5g
50g
Product Code. 11429436
Brand: Thermo Scientific Alfa Aesar A19146.09
Request Bulk or Custom Format
CAS: 21717-29-3 | C7H10N2 | 122.171 g/mol
The action of iodine on 2-amino-6-ethylpyridine in an alkaline medium gives 2-amino-5-iodo-6-ethylpyridine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
The action of iodine on 2-amino-6-ethylpyridine in an alkaline medium gives 2-amino-5-iodo-6-ethylpyridine.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 21717-29-3
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.171
MDL Number MFCD00218265
InChI Key JXKAUUVMXZIJNZ-UHFFFAOYSA-N
Synonym 2-amino-6-ethylpyridine, 6-ethyl-2-pyridinamine, 2-pyridinamine, 6-ethyl, 6-ethyl-2-pyridylamine, acmc-1clku, 2-pyridinamine,6-ethyl, 2-amino-6-ethyl-pyridine, 6-ethyl-pyridin-2-ylamine
PubChem CID 89021
IUPAC Name 6-ethylpyridin-2-amine
SMILES CCC1=NC(=CC=C1)N
Specifications
Odor Unpleasant
Refractive Index 1.568
Quantity 10 g
UN Number UN2810
Solubility Information Slightly soluble in water.
Formula Weight 122.17
Percent Purity 97%
Chemical Name or Material 2-Amino-6-ethylpyridine