Learn More
2-Amino-6-chloropurine, TRC
Product Code. 30179868
Change view
Click to view available options
Quantity:
1 g
5 g
50 g
Unit Size:
1g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179868 1 g 1g
30180284 50 g 50g
30179819 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30179868 Supplier Toronto Research Chemicals Supplier No. TRCA6035051G
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 10310-21-1
Molecular Formula C5 H4 Cl N5
Molecular Weight (g/mol) 169.57
Synonym 9H-Purin-2-amine, 6-chloro-, 1H-Purin-2-amine, 6-chloro- (9CI), Purine, 2-amino-6-chloro- (6CI,7CI,8CI), 6-Chloro-9H-purin-2-amine, 2-Amino-6-chloro-9H-purine, 2-Amino-6-chloropurine, 6-Chloro-1H-purin-2-amine, 6-Chloro-2-aminopurine, 6-Chloro-2-purinamine, 6-Chloro-7H-purin-2-amine, 6-Chloro-7H-purin-2-ylamine, 6-Chloro-9H-purin-2-yl-2-amine, 6-Chloroguanine, NSC 29570
IUPAC Name 6-chloro-9H-purin-2-amine
SMILES Nc1nc(Cl)c2nc[nH]c2n1
Specifications
Quantity 1 g
InChI Formula InChI=1S/C5H4ClN5/c6-3-2-4(9-1-8-2)11-5(7)10-3/h1H,(H3,7,8,9,10,11)
Formula Weight 169.0155
Chemical Name or Material 2-Amino-6-chloropurine