missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-5-nitro-4-picoline, 98%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
Specifications
Melting Point | 220.0°C to 225.0°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (GC) |
Quantity | 5 g |
Formula Weight | 153.14 |
Percent Purity | 98% |
Chemical Name or Material | 2-Amino-5-nitro-4-picoline |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'