Learn More
2-Amino-4-ethylpyridine, 97%
Product Code. 11453028
Change view
Click to view available options
Quantity:
2.5 g
10 g
Unit Size:
10g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11453028 2.5 g 2.5g
11463028 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11453028 Supplier Thermo Scientific Alfa Aesar Supplier No. A18754.88
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
CAS: 33252-32-3 | C7H11N2 | 123.18 g/mol
2-Amino-4-ethylpyridine is used as a precursor to prepare 1-ethyl-3-[5-(7-ethyl-imidazo[1,2-a]pyridin-2-yl)-2-hydroxy-phenyl]-urea. It is also used as an intermediate in organic synthesis. It is considered as an analogue of pirlimycin, which acts as an antibiotic.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Amino-4-ethylpyridine is used as a precursor to prepare 1-ethyl-3-[5-(7-ethyl-imidazo[1,2-a]pyridin-2-yl)-2-hydroxy-phenyl]-urea. It is also used as an intermediate in organic synthesis. It is considered as an analogue of pirlimycin, which acts as an antibiotic.
Solubility
Soluble in water, hexane, ethyl acetate and methyl tert-butyl ether.
Notes
Incompatible with strong oxidizing agents, strong acids and bases. Store in a cool place.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 33252-32-3
Molecular Formula C7H11N2
Molecular Weight (g/mol) 123.18
MDL Number MFCD00129025
InChI Key SJWHILBZPGQBJE-UHFFFAOYSA-O
Synonym 2-amino-4-ethylpyridine, 4-ethyl-pyridin-2-ylamine, 4-ethyl-2-pyridinamine, 2-pyridinamine, 4-ethyl, chembl62077, pubchem7108, 4-ethyl-2-pyridylamine, 4-ethyl-2-aminopyridine, acmc-20ao52
PubChem CID 118425
IUPAC Name 4-ethylpyridin-2-amine
SMILES CCC1=CC=[NH+]C(N)=C1
Specifications
Melting Point 65°C to 69°C
Quantity 2.5 g
UN Number UN2811
Solubility Information Soluble in water,hexane,ethyl acetate and methyl tert-butyl ether.
Formula Weight 122.17
Percent Purity 97%
Chemical Name or Material 2-Amino-4-ethylpyridine