Learn More
2-Amino-4,6-dimethoxypyrimidine, 98%, Thermo Scientific™
Product Code. 11420701
Change view
Click to view available options
Quantity:
2.5 g
10 g
Unit Size:
10g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11420701 10 g 10g
11410701 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11420701 Supplier Thermo Scientific Alfa Aesar Supplier No. B20152.09
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 36315-01-2
Molecular Formula C6H9N3O2
Molecular Weight (g/mol) 155.157
MDL Number MFCD00038832
InChI Key LVFRCHIUUKWBLR-UHFFFAOYSA-N
Synonym 2-amino-4,6-dimethoxypyrimidine, 2-pyrimidinamine, 4,6-dimethoxy, 4,6-dimethoxy-2-pyrimidinamine, 4,6-dimethoxy-2-aminopyrimidine, 4,6-dimethoxy-pyrimidin-2-ylamine, 2-amino-4,6-dimethoxy pyrimidine, 2-amino-4,6-dimethoxy-pyrimidine, 4,6-dimethoxypyrimidine-2-ylamine, maybridge1_002259, zlchem 350
PubChem CID 118946
IUPAC Name 4,6-dimethoxypyrimidin-2-amine
SMILES COC1=CC(=NC(=N1)N)OC
Specifications
Melting Point 94°C to 96°C
Quantity 10 g
Formula Weight 155.16
Percent Purity 98%
Chemical Name or Material 2-Amino-4,6-dimethoxypyrimidine