Learn More
2-Amino-3-iodopyridine, 98%
Product Code. 11494350
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11494350 1 g 1g
11404360 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11494350 Supplier Thermo Scientific Alfa Aesar Supplier No. H50001.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
CAS: 104830-06-0 | C5H5IN2 | 220.013 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 104830-06-0
Molecular Formula C5H5IN2
Molecular Weight (g/mol) 220.013
MDL Number MFCD01651885
InChI Key UUDNBWSHTUFGDQ-UHFFFAOYSA-N
Synonym 2-amino-3-iodopyridine, 3-iodopyrid-2-ylamine, 3-iodo-pyridin-2-ylamine, 3-iodo-2-pyridylamine, 2-pyridinamine, 3-iodo, iodo-aminopyridine, pubchem1265, 2-amino-3-iodpyridine, acmc-20a0df
PubChem CID 2763151
IUPAC Name 3-iodopyridin-2-amine
SMILES C1=CC(=C(N=C1)N)I
Spezifikation
Melting Point 88°C to 92°C
Quantity 1 g
Sensitivity Air and light sensitive
Formula Weight 220.01
Percent Purity 98%
Chemical Name or Material 2-Amino-3-iodopyridine