Learn More
2-Amino-1-methylimidazo[4,5-f]quinoline, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Amino-1-methylimidazo[4,5-F]quinoline |
| CAS | 102408-25-3 |
| Molecular Formula | C11 H10 N4 |
| Quantity | 10 mg |
| Synonym | 1H-Imidazo[4,5-f]quinolin-2-amine, 1-methyl-, 1-Methyl-1H-imidazo[4,5-f]quinolin-2-amine, 2-Amino-1-methylimidazo[4,5-f]quinoline |
| InChI Formula | InChI=1S/C11H10N4/c1-15-10-7-3-2-6-13-8(7)4-5-9(10)14-11(15)12/h2-6H,1H3,(H2,12,14) |
| SMILES | Cn1c(N)nc2ccc3ncccc3c12 |
| IUPAC Name | 1-methylimidazo[4,5-f]quinolin-2-amine |
| Molecular Weight (g/mol) | 198.22 |
| Formula Weight | 198.0905 |