Learn More
2-Allylphenol, 98+%
Product Code. p-7080980
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
Product Code. Quantity unitSize
11439642 100 g 100g
11449642 500 g 500g
2 options
Product Code. 11439642
Brand: Thermo Scientific Alfa Aesar A10755.22
Request Bulk or Custom Format
CAS: 1745-81-9 | C9H10O | 134.178 g/mol
2-Allylphenol is an protected intermediate in the synthesis of metabolites of Diclofenac (D436450), a nonsteroidal anti-inflammatory compound an decycloxygenase (COX) inhibitor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Allylphenol is an protected intermediate in the synthesis of metabolites of Diclofenac (D436450), a nonsteroidal anti-inflammatory compound an decycloxygenase (COX) inhibitor.
Solubility
Soluble in water(7 g/L).
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1745-81-9
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.178
MDL Number MFCD00002250
InChI Key QIRNGVVZBINFMX-UHFFFAOYSA-N
Synonym 2-allylphenol, o-allylphenol, phenol, o-allyl, phenol, 2-2-propenyl, phenol, 2-propenyl, 2-prop-2-en-1-yl phenol, 2-2-propenyl phenol, 2-allyl-phenol, unii-o04f145zjz, 2-prop-2-en-1-ylphenol
PubChem CID 15624
ChEBI CHEBI:39826
IUPAC Name 2-prop-2-enylphenol
SMILES C=CCC1=CC=CC=C1O
Specifications
Melting Point -6°C
Density 1.024
Boiling Point 220°C to 222°C
Flash Point 88°C (190°F)
Odor Phenol-like
Refractive Index 1.545
Quantity 100 g
UN Number UN2922
Beilstein 742121
Solubility Information Soluble in water(7g/L).
Formula Weight 134.18
Percent Purity ≥98%
Chemical Name or Material 2-Allylphenol