Learn More
Julabo™ 2-Adapters for Recirculating Cooler
Product Code. 11984708
Change view
Click to view available options
Connection Type:
G 1 1/4 in. F to 1 in. Barb. Fit
G 1 1/4 in. F to 3/4 in. Barb. Fit
G 1 1/4 in. F to NPT 1 in. M
G 1 1/4 in. F to NPT 1/2 in. M
G 1 1/4 in. F to NPT 3/4 in. M
G 1 1/4 in. Female to M16 x 1 Male
G 3/4 in. F to 1/2 in. Barb. Fit
G 3/4 in. F to NPT 1/2 in. M
G 3/4 in. F to NPT 3/4 in. M
G 3/4 in. Female to Barbed Fitting
G 3/4 in. Female to M16 x 1 Male
For Use With (Equipment):
FL11006,FL20006,FL2506,FL4006,FL7006
FL1203, FL1703, FL2503, FL4003
Unit Size:
Pack of 2
Product Code. Connection Type For Use With (Equipment) unitSize
11984708 G 1 1/4 in. F to 3/4 in. Barb. Fit FL11006,FL20006,FL2506,FL4006,FL7006 Pack of 2
11994708 G 1 1/4 in. F to 1 in. Barb. Fit FL11006,FL20006,FL2506,FL4006,FL7006 Pack of 2
11964708 G 1 1/4 in. Female to M16 x 1 Male FL11006,FL20006,FL2506,FL4006,FL7006 Pack of 2
11924718 G 1 1/4 in. F to NPT 1 in. M FL11006,FL20006,FL2506,FL4006,FL7006 Pack of 2
11914718 G 1 1/4 in. F to NPT 3/4 in. M FL11006,FL20006,FL2506,FL4006,FL7006 Pack of 2
11904718 G 1 1/4 in. F to NPT 1/2 in. M FL11006,FL20006,FL2506,FL4006,FL7006 Pack of 2
11924708 G 3/4 in. Female to Barbed Fitting FL1203, FL1703, FL2503, FL4003 Pack of 2
11984698 G 3/4 in. F to 1/2 in. Barb. Fit FL1203, FL1703, FL2503, FL4003 Pack of 2
11934708 G 3/4 in. F to NPT 1/2 in. M FL1203, FL1703, FL2503, FL4003 Pack of 2
11944708 G 3/4 in. F to NPT 3/4 in. M FL1203, FL1703, FL2503, FL4003 Pack of 2
11914708 G 3/4 in. Female to M16 x 1 Male FL1203, FL1703, FL2503, FL4003 Pack of 2
11 options
Product Code. 11984708
Brand: Julabo™ 8890045
Only null left
Specifications
For Use With (Equipment) FL11006,FL20006,FL2506,FL4006,FL7006
Type 2-Adapters
Connection Type G 1 1/4 in. F to 3/4 in. Barb. Fit