Learn More
Julabo™ 2-Adapters for Recirculating Cooler
Product Code. 11924718
G 1 1/4 in. F to NPT 1 in. M, FL11006,FL20006,FL2506,FL4006,FL7006, Pack of 2
Click to view available options
Connection Type:
G 1 1/4 in. F to 1 in. Barb. Fit
G 1 1/4 in. F to 3/4 in. Barb. Fit
G 1 1/4 in. F to NPT 1 in. M
G 1 1/4 in. F to NPT 1/2 in. M
G 1 1/4 in. F to NPT 3/4 in. M
G 1 1/4 in. Female to M16 x 1 Male
G 3/4 in. F to 1/2 in. Barb. Fit
G 3/4 in. F to NPT 1/2 in. M
G 3/4 in. F to NPT 3/4 in. M
G 3/4 in. Female to Barbed Fitting
G 3/4 in. Female to M16 x 1 Male
For Use With (Equipment):
FL11006,FL20006,FL2506,FL4006,FL7006
FL1203, FL1703, FL2503, FL4003
Unit Size:
Pack of 2
Product Code. 11924718
Brand: Julabo™ 8890051
Only undefined left
Only undefined left
Specifications
For Use With (Equipment) FL11006,FL20006,FL2506,FL4006,FL7006
Type 2-Adapters
Connection Type G 1 1/4 in. F to NPT 1 in. M
Product Content Correction
Product Title
Julabo™ 2-Adapters for Recirculating Cooler >