Learn More
2-Acetylanthracene, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Acetylanthracene |
| CAS | 10210-32-9 |
| Molecular Formula | C16 H12 O |
| Quantity | 5 g |
| InChI Formula | InChI=1S/C16H12O/c1-11(17)12-6-7-15-9-13-4-2-3-5-14(13)10-16(15)8-12/h2-10H,1H3 |
| SMILES | CC(=O)c1ccc2cc3ccccc3cc2c1 |
| IUPAC Name | 1-anthracen-2-ylethanone |
| Molecular Weight (g/mol) | 220.27 |
| Formula Weight | 220.0888 |