missing translation for 'onlineSavingsMsg'
Learn More

2-Acetyl-7-amino-1,2,3,4-tetrahydroisoquinoline, 95%, Thermo Scientific™

Product Code. p-7052527
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15499015 1 g 1g
15409025 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15499015 Supplier Thermo Scientific Alfa Aesar Supplier No. H35471.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Acetyl-7-amino-1,2,3,4-tetrahydroisoquinoline is used as an organic chemical synthesis intermediate.

Notes
Light sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81885-67-8
Molecular Formula C11H14N2O
Molecular Weight (g/mol) 190.246
MDL Number MFCD10016667
InChI Key XWVFMJLNNGXNSG-UHFFFAOYSA-N
Synonym 1-7-amino-3,4-dihydroisoquinolin-2 1h-yl ethanone, 2-acetyl-1,2,3,4-tetrahydroisoquinolin-7-amine, 2-acetyl-7-amino-1,2,3,4-tetrahydroisoquinoline, 7-isoquinolinamine, 2-acetyl-1,2,3,4-tetrahydro, 1-7-amino-3,4-dihydro-1h-isoquinolin-2-yl ethanone, n-acetyl-7-amino-1,2,3,4-tetrahydroisoquinoline, 1-7-amino-3,4-dihydro-2 1h-isoquinolinyl ethanone, 1-7-amino-3,4-dihydroisoquinolin-2 1h-yl ethan-1-one, ethanone, 1-7-amino-3,4-dihydro-2 1h-isoquinolinyl, 1-7-amino-1,2,3,4-tetrahydroisoquinolin-2-yl ethan-1-one
PubChem CID 12811352
IUPAC Name 1-(7-amino-3,4-dihydro-1H-isoquinolin-2-yl)ethanone
SMILES CC(=O)N1CCC2=C(C1)C=C(C=C2)N

Specifications

Melting Point 107°C to 111°C
Color Brown
Quantity 1 g
Sensitivity Light sensitive
Formula Weight 190.24
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Acetyl-7-amino-1,2,3,4-tetrahydroisoquinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.