missing translation for 'onlineSavingsMsg'
Learn More

2-Acetyl-5-chlorothiophene, 99%, Thermo Scientific™

Product Code. p-3756525 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
Product Code. Quantity Packaging unitSize
10147663 25 g Glass bottle 25g
1 options
This item is not returnable. View return policy

Product Code. 10147663

Brand: Thermo Scientific Acros 306700250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6310-09-4
Molecular Weight (g/mol) 160.62
MDL Number MFCD00005444
InChI Key HTZGPEHWQCRXGZ-UHFFFAOYSA-N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 43.0°C to 47.0°C
Color White to Yellow
Boiling Point 117.0°C to 118.0°C
Flash Point 108°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C6H5ClOS
Quantity 25 g
Synonym 2-bromothiophenol, o-bromothiophenol, o-bromobenzenethiol, 2-bromo thiophenol, benzenethiol, 2-bromo, benzenethiol, o-bromo, 2-bromobenzene-1-thiol, 2-bromo-benzenethiol, 2-bromo-thiophenol, 2-bromobenzene thiol
SMILES CC(=O)C1=CC=C(Cl)S1
IUPAC Name 2-bromobenzenethiol
PubChem CID 80598
Formula Weight 160.62
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2-Acetyl-5-chlorothiophene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.