Learn More
2,6-Dimethylpyridine, Sigma-Aldrich™
Specifications
Specifications
| CAS | 108-48-5 |
| Melting Point | -6°C |
| Density | 0.923 g/cm3 at 20°C |
| Boiling Point | 144°C/1013 hPa |
| Molecular Formula | 2,6-(CH3)2C5H3N |
| MDL Number | MFCD00006345 |
| Quantity | 250 mL |
| Synonym | 2,6-Dimethylpyridine, 2,6-Lutidine, 2,6-Dimethylpyridine |
| Molecular Weight (g/mol) | 107.15 |
| Grade | Research |