Learn More
2,6-Dimethylaniline, Sigma-Aldrich™
Product Code. 15829680
Change view
Click to view available options
Quantity:
5 mL
250 mL
1 L
Unit Size:
1 L
250mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15829680 1 L 1 L
15809680 5 mL 5mL
15819680 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15829680 Supplier Sigma Aldrich Supplier No. 8086711000
 Restricted Item
Estimated Shipment: 19-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 87-62-7
Grade Research
Molecular Formula 2,6-(CH3)2C6H3NH2
Molecular Weight (g/mol) 121.18
Specifications
Melting Point 10°C to 12°C
Density 0.98 g/cm3 at 20°C
Quantity 1 L
MDL Number MFCD00007747
Synonym 2,6-Dimethylaniline, vic.-m-Xylidine, 2,6-Dimethylaniline
Boiling Point 216°C/1013 hPa
Chemical Name or Material 2,6-Dimethylaniline