Learn More
2,6-Difluoroaniline, 98%, Thermo Scientific Chemicals
Catalog Number p-7020907
Click to view available options
Quantity:
5 g
25 g
100 g
5509-65-9
C6H5F2N
129.11
MFCD00007655
ODUZJBKKYBQIBX-UHFFFAOYSA-N
benzenamine, 2,6-difluoro, 2,6-difluoro-phenylamine, 2,6-difluoro aniline, 2,6-difluorobenzenamine, 2,6-difluoranilin, 2,6 difluoroaniline, 2,6-diflouroaniline, pubchem4034, 2,6-difuluoroaniline
79647
2,6-difluoroaniline
NC1=C(F)C=CC=C1F
5509-65-9
C6H5F2N
129.11
MFCD00007655
ODUZJBKKYBQIBX-UHFFFAOYSA-N
benzenamine, 2,6-difluoro, 2,6-difluoro-phenylamine, 2,6-difluoro aniline, 2,6-difluorobenzenamine, 2,6-difluoranilin, 2,6 difluoroaniline, 2,6-diflouroaniline, pubchem4034, 2,6-difuluoroaniline
79647
2,6-difluoroaniline
NC1=C(F)C=CC=C1F
2,6-Difluoroaniline is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,6-Difluoroaniline is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Incompatible to acid chlorides, acid anhydrides, oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 1.275
Boiling Point 152°C to 154°C
Flash Point 51°C (124°F)
Odor Characteristic
Refractive Index 1.508
Quantity 5 g
UN Number UN1992
Beilstein 2802697
Solubility Information Slightly soluble in water.
Formula Weight 129.11
Percent Purity 98%
Chemical Name or Material 2,6-Difluoroaniline