Learn More
2,6-Difluoro-4-hydroxybenzoic acid, 95%
Product Code. 11302139
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
11302139 1 g 1g
15433407 250 mg 250mg
2 options
Product Code. 11302139
Brand: Thermo Scientific Alfa Aesar H26134.03
Request Bulk or Custom Format
CAS: 214917-68-7 | C7H4F2O3 | 174.10 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 214917-68-7
Molecular Formula C7H4F2O3
Molecular Weight (g/mol) 174.10
MDL Number MFCD03094500
InChI Key NFIQGYBXSJQLSR-UHFFFAOYSA-N
Synonym 2,6-difluoro-4-hydroxy-benzoic acid, benzoic acid, 2,6-difluoro-4-hydroxy, pubchem4938, acmc-1cles, ksc495o6f, 4-carboxy-3,5-difluorophenol
PubChem CID 2778776
IUPAC Name 2,6-difluoro-4-hydroxybenzoic acid
SMILES OC(=O)C1=C(F)C=C(O)C=C1F
Specifications
Melting Point 170°C to 171°C (decomposition)
Quantity 1 g
Formula Weight 174.1
Percent Purity 95%
Chemical Name or Material 2,6-Difluoro-4-hydroxybenzoic acid