Learn More
2,6-Di-tert-butyl-4-methylphenol, Sigma-Aldrich™
Product Code. 15891611
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
25 kg
Unit Size:
100g
2.5kg
25kg
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15891611 25 kg 25kg
15876690 100 g 100g
15871611 500 g 500g
15881611 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15891611 Supplier Sigma Aldrich Supplier No. 8220219025
 Restricted Item
Estimated Shipment: 18-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 128-37-0
Grade Research
Molecular Weight (g/mol) 220.35
Specifications
Melting Point 69°C to 70°C
Density 1.05 g/cm3 at 20°C
Boiling Point 265°C/1013 hPa
MDL Number MFCD00011644
Quantity 25 kg
Synonym 2,6-Di-tert-butyl-4-methylphenol, 2,6-Di-tert-butyl-4-methylphenol