Learn More
2,6-Di-tert-butyl-4-hydroxymethylphenol, Mikromol™
Product Code. 17959364
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17959364 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17959364 Supplier Mikromol Supplier No. MM0679.010025
 Restricted Item
Estimated Shipment: 16-07-2026
Add to basket
Add to basket
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2,6-Di-tert-butyl-4-hydroxymethylphenol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2,6-Di-tert-butyl-4-hydroxymethylphenol
CAS 88-26-6
Analyte or Component Names 2,6-Di-tert-butyl-4-hydroxymethylphenol
Shipping Condition Room Temperature
Molecular Formula C15 H24 O2
Certifications/Compliance ISO 9001
Synonym Benzenemethanol, 3,5-bis(1,1-dimethylethyl)-4-hydroxy-, 2,6-Di-tert-butyl-4-hydroxymethylphenol, Benzyl alcohol, 3,5-di-tert-butyl-4-hydroxy- (6CI,7CI,8CI), 3,5-Bis(1,1-dimethylethyl)-4-hydroxybenzenemethanol, 2,6-Di-tert-butyl-4-hydroxymethylphenol, 3,5-Di-t-butyl-4-hidroxy-benzyl alcohol, 3,5-Di-tert-butyl-4-hydroxybenzyl alcohol, 3,5-Di-tert-butyl-4-hydroxyphenylmethanol, 4-Hydroxy-3,5-di-tert-butylbenzyl alcohol, 4-Hydroxymethyl-2,6-di-tert-butylphenol, AO 754, Ethanox 754, Ionox 100, NSC 159133
InChI Formula InChI=1S/C15H24O2/c1-14(2,3)11-7-10(9-16)8-12(13(11)17)15(4,5)6/h7-8,16-17H,9H2,1-6H3
SMILES CC(C)(C)c1cc(CO)cc(c1O)C(C)(C)C
IUPAC Name 2,6-ditert-butyl-4-(hydroxymethyl)phenol
Molecular Weight (g/mol) 236.35
Quantity 25 mg
Formula Weight 236.1776 g/mol