missing translation for 'onlineSavingsMsg'
Learn More

Chem-Lab 2.5 l Hexane-(n) a.r. - 99+% C6H14

Brand:  Chem-Lab CL00.0802.2500


2.5 l Hexane-(n) a.r. - 99+% C6H14

Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 16082002

  • 1560.00 NOK / 2.5 L

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Hexane-(n)

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H361f-Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P331-Do NOT induce vomiting.
  • P403+P235-Store in a well-ventilated place. Keep cool.

Supplemental information
  • MIXTURE LIST-Contain: Hexane-(n)

SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.