Learn More
2,5-Dimethylcinnamic acid, 98+%, Thermo Scientific™
Click to view available options
Quantity:
2 g
10 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
2,5-Dimethylcinnamic acid, is used for organic synthesize intermediate
Solubility
Insoluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
2,5-Dimethylcinnamic acid, is used for organic synthesize intermediate
Solubility
Insoluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Melting Point | 129°C to 131°C |
Quantity | 2 g |
Beilstein | 3244516 |
Solubility Information | Insoluble in water. |
Formula Weight | 176.22 |
Percent Purity | ≥98% |
Chemical Name or Material | 2,5-Dimethylcinnamic acid |