Learn More
(2,5-Dimethoxyphenyl)boronic acid, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 107099-99-0 |
| Molecular Formula | C8H11BO4 |
| MDL Number | MFCD01318181 |
| Quantity | 500 g |
| SMILES | COC1=CC(B(O)O)=C(OC)C=C1 |
| Molecular Weight (g/mol) | 181.98 |
| Percent Purity | 97% |
| Chemical Name or Material | (2,5-Dimethoxyphenyl)boronic acid |