missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluorophenylhydrazine hydrochloride, 99%, Thermo Scientific™

Catalog Number 11388385
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
175135-73-6
C6H7ClF2N2
180.583
MFCD00013385
LZKLZCSHMDRKJH-UHFFFAOYSA-N
2,5-difluorophenylhydrazine hydrochloride, 2,5-difluorophenyl hydrazine hydrochloride, 2,5-difluorophenylhydrazine hcl, 2,5-difluoro-phenyl-hydrazine hydrochloride, hydrazine, 2,5-difluorophenyl-, monohydrochloride, 2,5-difluorphenylhydrazine hydrochloride, 2,5-difluorophenylhy, pubchem1082, acmc-1c9k4, ksc491e8d
24720967
(2,5-difluorophenyl)hydrazine;hydrochloride
C1=CC(=C(C=C1F)NN)F.Cl
This item is not returnable. View return policy
175135-73-6
C6H7ClF2N2
180.583
MFCD00013385
LZKLZCSHMDRKJH-UHFFFAOYSA-N
2,5-difluorophenylhydrazine hydrochloride, 2,5-difluorophenyl hydrazine hydrochloride, 2,5-difluorophenylhydrazine hcl, 2,5-difluoro-phenyl-hydrazine hydrochloride, hydrazine, 2,5-difluorophenyl-, monohydrochloride, 2,5-difluorphenylhydrazine hydrochloride, 2,5-difluorophenylhy, pubchem1082, acmc-1c9k4, ksc491e8d
24720967
(2,5-difluorophenyl)hydrazine;hydrochloride
C1=CC(=C(C=C1F)NN)F.Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point ∽211°C (decomposition)
Quantity 1 g
Beilstein 8640307
Formula Weight 180.59
Percent Purity 99%
Chemical Name or Material 2,5-Difluorophenylhydrazine hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'