Learn More
2,5-Difluorophenylacetonitrile, 97%, Thermo Scientific™
Product Code. 11489716
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489716 1 g 1g
11499716 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11489716 Supplier Thermo Scientific Alfa Aesar Supplier No. B20246.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 69584-87-8
Molecular Formula C8H5F2N
Molecular Weight (g/mol) 153.13
MDL Number MFCD00009972
InChI Key UIMMFRUOZOWROM-UHFFFAOYSA-N
Synonym 2,5-difluorophenylacetonitrile, 2,5-difluorobenzyl cyanide, 2-2,5-difluorophenyl acetonitrile, 2,5-difluorophenyl acetonitrile, benzeneacetonitrile, 2,5-difluoro, 2,5-difluoro-phenyl-acetonitrile, 2-2,5-difluorophenyl ethanenitrile, 2,5-difluorophenylacetonitirle, pubchem4097, 2,5-difluorobenzylcyanide
PubChem CID 592780
IUPAC Name 2-(2,5-difluorophenyl)acetonitrile
SMILES FC1=CC=C(F)C(CC#N)=C1
Specifications
Density 1.233
Boiling Point 90°C (5 mmHg)
Flash Point 103°C (217°F)
Refractive Index 1.4834
Quantity 1 g
UN Number UN3276
Beilstein 2576318
Formula Weight 153.13
Percent Purity 97%
Chemical Name or Material 2,5-Difluorophenylacetonitrile