Learn More
2,5-Dichloronitrobenzene, Mikromol™
Product Code. 17981916
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17981916 100 mg 100mg
30052431 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17981916 Supplier Mikromol Supplier No. MM1273.220100
 Restricted Item
Estimated Shipment: 20-07-2026
Add to basket
Add to basket
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2,5-Dichloronitrobenzene
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2,5-Dichloronitrobenzene
CAS 89-61-2
Analyte or Component Names 2,5-Dichloronitrobenzene
Shipping Condition Room Temperature
Molecular Formula C6 H3 Cl2 N O2
Certifications/Compliance ISO 17025
Synonym Benzene, 1,4-dichloro-2-nitro-, Benzene, 1,4-dichloro-3-nitro- (1CI), 1,4-Dichloro-2-nitrobenzene, 1,4-Dichloro-3-nitrobenzene, 1-Nitro-2,5-dichlorobenzene, 2,5-Dichloro-1-nitrobenzene, 2,5-Dichloronitrobenzene, 2-Nitro-1,4-dichlorobenzene, NSC 406125, NSC 6294, Nitro-p-dichlorobenzene
InChI Formula InChI=1S/C6H3Cl2NO2/c7-4-1-2-5(8)6(3-4)9(10)11/h1-3H
SMILES [O-][N+](=O)c1cc(Cl)ccc1Cl
IUPAC Name 1,4-dichloro-2-nitrobenzene
Molecular Weight (g/mol) 192
Quantity 100 mg
Formula Weight 190.9541 g/mol