Learn More
2,5-Dibromobenzoic acid, 98%
Product Code. 11467377
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 11467377
Brand: Thermo Scientific Alfa Aesar A15077.14
Request Bulk or Custom Format
CAS: 610-71-9 | C7H4Br2O2 | 279.92 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 610-71-9
Molecular Formula C7H4Br2O2
Molecular Weight (g/mol) 279.92
MDL Number MFCD00016494
InChI Key SQQKOTVDGCJJKI-UHFFFAOYSA-N
Synonym benzoic acid, 2,5-dibromo, rarechem fh 1w 0049, pubchem3859, 2,5-dibromobenzoicacid, 2,5-dibromo-benzoic acid, benzoicacid, 2,5-dibromo, 4-09-00-01027 beilstein handbook reference, ksc353s4r, acmc-1b114, 2,5-dibromobenzoic acid
PubChem CID 11891
IUPAC Name 2,5-dibromobenzoic acid
SMILES OC(=O)C1=CC(Br)=CC=C1Br
Specifications
Melting Point 153°C to 157°C
Quantity 25 g
Beilstein 1868193
Formula Weight 279.93
Percent Purity 98%
Chemical Name or Material 2,5-Dibromobenzoic acid