missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromo-3-methylpyridine, 98%, Thermo Scientific™

Catalog Number 11416260
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
3430-18-0
C6H5Br2N
250.92
MFCD02093085
LIMXEVCFAUTBCK-UHFFFAOYSA-N
2,5-dibromo-3-picoline, 2,5-dibromo-3-methyl-pyridine, pyridine, 2,5-dibromo-3-methyl, 2,5-dibromo-3-methylpyridine, 2,5-dibrom-3-methylpyridin, pubchem1138, acmc-209i6e, ksc495g9h, 2,5-dibromo-3-methyl pyridine
817680
2,5-dibromo-3-methylpyridine
CC1=CC(Br)=CN=C1Br
This item is not returnable. View return policy
3430-18-0
C6H5Br2N
250.92
MFCD02093085
LIMXEVCFAUTBCK-UHFFFAOYSA-N
2,5-dibromo-3-picoline, 2,5-dibromo-3-methyl-pyridine, pyridine, 2,5-dibromo-3-methyl, 2,5-dibromo-3-methylpyridine, 2,5-dibrom-3-methylpyridin, pubchem1138, acmc-209i6e, ksc495g9h, 2,5-dibromo-3-methyl pyridine
817680
2,5-dibromo-3-methylpyridine
CC1=CC(Br)=CN=C1Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 43°C to 47°C
Flash Point >110°C (230°F)
Quantity 1 g
UN Number UN2811
Formula Weight 250.92
Percent Purity 98%
Chemical Name or Material 2,5-Dibromo-3-methylpyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'