Learn More
2,5-Di-tert-butylhydroquinone, 98+%
Product Code. 11482038
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
Product Code. Quantity unitSize
11482038 250 g 250g
11472038 1000 g 1000g
2 options
Product Code. 11482038
Brand: Thermo Scientific Alfa Aesar A14606.30
Request Bulk or Custom Format
CAS: 88-58-4 | C14H22O2 | 222.33 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 88-58-4
Molecular Formula C14H22O2
Molecular Weight (g/mol) 222.33
MDL Number MFCD00008825
InChI Key JZODKRWQWUWGCD-UHFFFAOYSA-N
Synonym 2,5-di-tert-butylhydroquinone, 2,5-di-tert-butylbenzene-1,4-diol, dibug, dtbhq, dybug, santovar o, di-t-butylhydroquinone, 2,5-di-tert-butylquinol, nonflex alba, 2,5-di-t-butylhydroquinone
PubChem CID 2374
ChEBI CHEBI:41094
IUPAC Name 2,5-ditert-butylbenzene-1,4-diol
SMILES CC(C)(C)C1=CC(O)=C(C=C1O)C(C)(C)C
Specifications
Melting Point 214°C to 218°C
Density 1.07
Boiling Point 321°C
Flash Point >200°C (392°F)
Quantity 250 g
Beilstein 2049542
Formula Weight 222.33
Percent Purity ≥98%
Chemical Name or Material 2,5-Di-tert-butylhydroquinone
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : 2,5-Di-tert-butylhydroquinone