Learn More
2-(4-Morpholinyl)ethyl isothiocyanate, 97%, Thermo Scientific Chemicals
Catalog Number p-7045807
Click to view available options
Quantity:
2 g
10 g
63224-35-1
C7H12N2OS
172.25
MFCD00041221
SDNSCXCXVQHBGH-UHFFFAOYSA-N
2-morpholinoethyl isothiocyanate, 4-2-isothiocyanatoethyl morpholine, 2-4-morpholino ethyl isothiocyanate, 2-morpholinoethylisothiocyanate, 4-2-isothiocyanato-ethyl-morpholine, 2-morpholin-4-ylethanisothiocyanate, 2-4-morpholinyl ethyl isothiocyanate, 4-2-isothiocyanatoethyl morpholine #
143957
4-(2-isothiocyanatoethyl)morpholine
S=C=NCCN1CCOCC1
63224-35-1
C7H12N2OS
172.25
MFCD00041221
SDNSCXCXVQHBGH-UHFFFAOYSA-N
2-morpholinoethyl isothiocyanate, 4-2-isothiocyanatoethyl morpholine, 2-4-morpholino ethyl isothiocyanate, 2-morpholinoethylisothiocyanate, 4-2-isothiocyanato-ethyl-morpholine, 2-morpholin-4-ylethanisothiocyanate, 2-4-morpholinyl ethyl isothiocyanate, 4-2-isothiocyanatoethyl morpholine #
143957
4-(2-isothiocyanatoethyl)morpholine
S=C=NCCN1CCOCC1
2-(4-Morpholinyl)ethyl isothiocyanate is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-(4-Morpholinyl)ethyl isothiocyanate is used as pharmaceutical intermediates.
Solubility
Hydrolyzes in water.
Notes
Store at 4°C. Store under dry inert gas. Protect from humidity and water. Incompatible with moisture, amines, alcohols and oxidizing agents. Protect from heat. Moisture Sensitive.
TRUSTED_SUSTAINABILITY
Density 1.11
Boiling Point 162°C to 163°C (10 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.539
Quantity 2 g
UN Number UN2922
Beilstein 1210530
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes in water.
Formula Weight 172.25
Percent Purity 97%
Chemical Name or Material 2-(4-Morpholinyl)ethyl isothiocyanate