missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Methylphenyl)ethanol, 99%

Product Code. 11439956
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. Quantity unitSize
11439956 1 g 1g
11449956 5 g 5g
11459956 25 g 25g
3 options
This item is not returnable. View return policy

Product Code. 11439956

Brand: Thermo Scientific Alfa Aesar B20918.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 699-02-5 | C9H12O | 136.194 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 699-02-5
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00002904
InChI Key DAVFJRVIVZOKKS-UHFFFAOYSA-N
Synonym 2-4-methylphenyl ethanol, 2-p-tolyl ethanol, 4-methylphenethyl alcohol, benzeneethanol, 4-methyl, 2-p-tolylethanol, p-methylphenethyl alcohol, 4-methylbenzeneethanol, 2-4-methylphenyl ethan-1-ol, p-methylphenylethyl alcohol, 2-p-methylphenyl ethanol
PubChem CID 95294
IUPAC Name 2-(4-methylphenyl)ethanol
SMILES CC1=CC=C(C=C1)CCO

Specifications

Density 0.98
Boiling Point 244°C to 245°C
Flash Point 107°C (225°F)
Refractive Index 1.526
Quantity 1 g
Formula Weight 136.2
Percent Purity 99%
Chemical Name or Material 2-(4-Methylphenyl)ethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.