missing translation for 'onlineSavingsMsg'
Learn More

2,4-Hexadiene, tech. 90%, mixture of isomers, Thermo Scientific™

Catalog Number p-7035681
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
592-46-1
C6H10
82.15
MFCD00009297,MFCD00009297
APPOKADJQUIAHP-CIIODKQPSA-N
unii-g4m9133vs9, 2,4-hexadiene, e,z, cis,trans-2,4-hexadiene, 2,4-hexadiene, e , z-ch3ch=chch=chch3, cis,trans-hexa-2,4-diene, 2,4-hexadiene, 2e,4z, cis-2-trans-4-hexadiene, e,z-2,4-hexadiene, z,e-2,4-hexadiene
643786
(2Z,4E)-hexa-2,4-diene
C\C=C\C=C/C
This item is not returnable. View return policy
592-46-1
C6H10
82.15
MFCD00009297,MFCD00009297
APPOKADJQUIAHP-CIIODKQPSA-N
unii-g4m9133vs9, 2,4-hexadiene, e,z, cis,trans-2,4-hexadiene, 2,4-hexadiene, e , z-ch3ch=chch=chch3, cis,trans-hexa-2,4-diene, 2,4-hexadiene, 2e,4z, cis-2-trans-4-hexadiene, e,z-2,4-hexadiene, z,e-2,4-hexadiene
643786
(2Z,4E)-hexa-2,4-diene
C\C=C\C=C/C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2,4-Hexadiene is used in the production of 3,6-dimethyl-3,6-dihydro-[1,2]dithiine.

Solubility
Miscible with ethanol, ethyl ether and chloroform. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 0.72
Boiling Point 82°C
Flash Point −14°C (−7°F)
Linear Formula CH3CH=CHCH=CHCH3
Refractive Index 1.445
Quantity 1 g
UN Number UN2458
Solubility Information Miscible with ethanol,ethyl ether and chloroform. Immiscible with water.
Formula Weight 82.15
Percent Purity 90%
Grade Technical
Chemical Name or Material 2,4-Hexadiene, mixture of isomers
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'