Learn More
2,4-Dioxo-1,2,3,4-tetrahydropyrimidine-5-sulfonyl chloride, 97%
Product Code. 15451156
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15451156 5 g 5g
15461156 250 mg 250mg
15441156 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15451156 Supplier Thermo Scientific Alfa Aesar Supplier No. H54410.06
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
CAS: 28485-18-9 | C4H3ClN2O4S | 210.59 g/mol
2,4-Dioxo-1,2,3,4-tetrahydropyrimidine-5-sulfonyl chloride is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,4-Dioxo-1,2,3,4-tetrahydropyrimidine-5-sulfonyl chloride is used as pharmaceutical intermediate.
Notes
Moisture sensitive. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 28485-18-9
Molecular Formula C4H3ClN2O4S
Molecular Weight (g/mol) 210.59
MDL Number MFCD06655485
InChI Key FQHNJCWWBKYCJW-UHFFFAOYSA-N
Synonym 2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-sulfonyl chloride, 2,4-dioxo-1,3-dihydropyrimidine-5-sulfonyl chloride, 1,2,3,4-tetrahydro-2,4-dioxo-5-pyrimidinesulfonyl chloride, 5-pyrimidinesulfonylchloride, 1,2,3,4-tetrahydro-2,4-dioxo, uracil-5-sulphonyl chloride, 5-chlorosulfonyl-1,3-dihydropyrimidine-2,4-dione, 2,4-dioxo-1,2,3,4-tetrahydro-5-pyrimidinesulfonyl chloride, 2,4-dioxo-1,2,3,4-tetrahydro-pyrimidine-5-sulfonyl chloride
PubChem CID 234620
SMILES ClS(=O)(=O)C1=CNC(=O)NC1=O
Specifications
Quantity 5 g
UN Number UN3261
Sensitivity Moisture sensitive
IUPAC Name 2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-sulfonyl chloride
Formula Weight 209.95
Percent Purity 97%
Chemical Name or Material 2,4-Dioxo-1,2,3,4-tetrahydropyrimidine-5-sulfonyl chloride