Learn More
2,4-Dimethylbenzoyl chloride, 97%, Thermo Scientific™
Product Code. 11325379
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11325379 5 g 5g
11315379 1 g 1g
2 options
Product Code. 11325379
Brand: Thermo Scientific Alfa Aesar H50387.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 21900-42-5
Molecular Formula C9H9ClO
Molecular Weight (g/mol) 168.62
MDL Number MFCD00798005
InChI Key JUOGRBULLOEKKQ-UHFFFAOYSA-N
Synonym benzoyl chloride, 2,4-dimethyl, 2,4-dimethyl-benzoyl chloride, benzoyl chloride,2,4-dimethyl, acmc-20aocb, 2.4-dimethylbenzoylchlorid, 2,4-di-methylbenzoyl chloride, 2,4-dimethyl benzoyl chloride, buttpark 48\06-96, 2,4-dimethyl-1-benzenecarbonyl chloride, benzoyl chloride, 2,4-dimethyl-6ci,8ci,9ci
PubChem CID 140867
IUPAC Name 2,4-dimethylbenzoyl chloride
SMILES CC1=CC(=C(C=C1)C(=O)Cl)C
Specifications
Melting Point 24°C to 26°C
Boiling Point 82°C (1 mmHg)
Quantity 5 g
UN Number UN3265
Formula Weight 168.62
Percent Purity 97%
Chemical Name or Material 2,4-Dimethylbenzoyl chloride