Learn More
2',4'-Dimethylacetophenone, TRC
Specifications
Specifications
| Chemical Name or Material | 2',4'-Dimethylacetophenone |
| CAS | 89-74-7 |
| Molecular Formula | C10H12O |
| Quantity | 1 g |
| Synonym | 1-(2,4-Dimethylphenyl)Ethanone, 2,4-Dimethylacetophenone, 2,4-Dimethylphenyl Methyl Ketone, 2',4'-Dimethylacetophenone |
| InChI Formula | InChI=1S/C10H12O/c1-7-4-5-10(9(3)11)8(2)6-7/h4-6H,1-3H3 |
| SMILES | CC(=O)c1ccc(C)cc1C |
| Molecular Weight (g/mol) | 148.2 |
| Formula Weight | 148.0888 |