Learn More
2,4-Dimethyl-1,3-pentadiene, 98%
Product Code. p-3736334 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
Product Code. Quantity Packaging unitSize
10684822 5 g Glass bottle 5g
10616362 25 g Glass bottle 25g
10571974 100 g Glass bottle 100g
3 options
Product Code. 10684822
Brand: Thermo Scientific Acros 155360050
Request Bulk or Custom Format
CAS: 1000-86-8 | C7H12 | 96.17 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1000-86-8
Molecular Formula C7H12
Molecular Weight (g/mol) 96.17
MDL Number MFCD00008903
InChI Key CMSUNVGIWAFNBG-UHFFFAOYSA-N
Synonym 2,4-dimethyl-1,3-pentadiene, 1,1,3-trimethylbutadiene, 1,3-pentadiene, 2,4-dimethyl, acmc-20aoci, 1,3-trimethylbutadiene, 1,3-pentadiene,2,4-dimethyl, cmsunvgiwafnbg-uhfffaoysa, ch3 2c=chc ch3 =ch2, 1,3-pentadiene, 2,4-dimethyl-8ci 9ci
PubChem CID 66080
IUPAC Name 2,4-dimethylpenta-1,3-diene
SMILES CC(=CC(=C)C)C
Specifications
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.7400g/mL
Boiling Point 94°C
Flash Point 10°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (CH3)2C=CHC(CH3)=CH2
Refractive Index 1.4400 to 1.442
Quantity 5 g
Beilstein 01, 257
Specific Gravity 0.74
Formula Weight 96.17
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2, 4-Dimethyl-1, 3-pentadiene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,4-Dimethyl-1,3-pentadiene
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.