missing translation for 'onlineSavingsMsg'
Learn More

2',4'-Dimethoxyacetophenone, 98%

Product Code. p-7022972
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11438566 25 g 25g
11448566 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11438566 Supplier Thermo Scientific Alfa Aesar Supplier No. A15982.14
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 829-20-9 | C10H12O3 | 180.203 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 829-20-9
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00008727
InChI Key VQTDPCRSXHFMOL-UHFFFAOYSA-N
Synonym 2',4'-dimethoxyacetophenone, 1-2,4-dimethoxyphenyl ethanone, 2,4-dimethoxyacetophenone, 1-2,4-dimethoxyphenyl ethan-1-one, ethanone, 1-2,4-dimethoxyphenyl, acetophenone, 2',4'-dimethoxy, peonol methyl ether, 1-acetyl-2,4-dimethoxybenzene, pubchem10582, acmc-209pqs
PubChem CID 70016
IUPAC Name 1-(2,4-dimethoxyphenyl)ethanone
SMILES CC(=O)C1=C(C=C(C=C1)OC)OC

Specifications

Melting Point 39°C to 41°C
Boiling Point 287°C to 289°C
Flash Point >110°C (230°F)
Quantity 25 g
Sensitivity Light sensitive
Formula Weight 180.2
Percent Purity 98%
Chemical Name or Material 2',4'-Dimethoxyacetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.